missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
TROY/TNFRSF19 Monoclonal specifically detects TROY/TNFRSF19 in Mouse samples. It is validated for ELISA.
Especificaciones
Especificaciones
| Antígeno | TROY/TNFRSF19 |
| Aplicaciones | ELISA |
| Clasificación | Monoclonal |
| Clon | 208 |
| Conjugado | Unconjugated |
| Dilución | ELISA 1:5000-1:10000 |
| Formulación | 0.2 um filtered solution in PBS |
| Alias de gen | TAJ-alpha, TAJToxicity and JNK inducer, TRADE, TROYtumor necrosis factor receptor superfamily member 19, tumor necrosis factor receptor superfamily, member 19 |
| Especie del huésped | Rabbit |
| Inmunógeno | Recombinant Mouse TROY/TNFRSF19 protein |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?