Learn More
TRIM17 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 13663-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies. The protein is expressed almost exclusively in the testis, but its function is unknown. Multiple alternatively spliced transcript variants have been found for this gene.Especificaciones
| TRIM17 | |
| Polyclonal | |
| Unconjugated | |
| TRIM17 | |
| RBCC, RING finger protein 16, RNF16, terf, Testis RING finger protein, TRIM17, tripartite motif containing 17 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 51127, 56631, 64702 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q7TPM3, Q9WV59, Q9Y577 | |
| TRIM17 | |
| TRIM17 Fusion Protein Ag4572 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |