missing translation for 'onlineSavingsMsg'
Learn More

TRIM16 Antibody, Novus Biologicals™

Código de producto. 18226234 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μL
25 μL
Tamaño de la unidad:
100 microlitros
25 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18226234 100 μL 100 microlitros
18643718 25 μL 25 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18226234 Supplier Novus Biologicals Supplier No. NBP256990

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TRIM16 Polyclonal specifically detects TRIM16 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno TRIM16
Aplicaciones Immunocytochemistry, Immunofluorescence
Clasificación Polyclonal
Conjugado Unconjugated
Dilución Immunocytochemistry/Immunofluorescence 1-4 ug/ml
Formulación PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Alias de gen DKFZp434O1826, EBBPtripartite motif-containing 16, Estrogen-responsive B box protein, tripartite motif containing 16, tripartite motif-containing protein 16
Símbolos de los genes TRIM16
Especie del huésped Rabbit
Inmunógeno This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:YCKFKNTEDITFPSVYIGLKDKLSGIRKVITESTVHLIQLLENYKKKLQEFSKEEEYDIRTQVSAIVQRKYWTSKPEPST
Método de purificación Affinity Purified
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 10626
Especificidad de la prueba Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Especies diana Human
Contenido y almacenamiento Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.