missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRHR Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP2-24726
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
TRHR Polyclonal specifically detects TRHR in Human, Canine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones
| TRHR | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.2% Gelatin with 0.05% Sodium Azide | |
| TRHR | |
| A portion of amino acids 350-400 of human TRH-R protein was used as the immunogen. | |
| 0.1 mg | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee, rat, mouse and cow and 76% homologous in wild boar. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Western Blot 0.5-1 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml | |
| MGC141920, Thyroliberin receptor, thyrotropin-releasing hormone receptor, TRH-R, TRHR1 | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| 7201 | |
| Human, Canine, Primate | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur