missing translation for 'onlineSavingsMsg'
Learn More

TRF-2 Antibody (4A794.15) - BSA Free, Novus Biologicals™

Código de producto. p-4997292 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.025 mg
0.1 mg
Tamaño de la unidad:
0.10 miligramo
25 microgramos
Código de producto. Cantidad unitSize
18790364 0.025 mg 25 microgramos
18765093 0.1 mg 0.10 miligramo
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 18790364

Marca: Novus Biologicals NB10056506SS

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

TRF-2 Monoclonal specifically detects TRF-2 in Human, Mouse, Rat, Deer, Marsupial samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Proximity Ligation Assay, CyTOF-ready.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno TRF-2
Aplicaciones Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry
Clasificación Monoclonal
Clon 4A794.15
Concentración 1 mg/mL
Conjugado Unconjugated
Dilución Western Blot 2-4 ug/ml, Simple Western 1:50, Flow Cytometry 0.1 ug/10^6 cells, ELISA 2 ug/ml, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 2 ug/10^6 cells, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 5 ug/ml, Immunoblotting reported in scientific literature (PMID 23708666), Proximity Ligation Assay reported in scientific literature (PMID 27366950), Chromatin Immunoprecipitation (ChIP) 1:10-1:500, CyTOF-ready
N.º de referencia del gen Q15554
Alias de gen 4A794, 4A794 trf2, 4A794.15, 4A794.15 trf2, anti-trf2 4A794, anti-trf2 4A794.15, clone 4A794, clone 4A794.15, Telomeric DNA-binding protein, telomeric repeat binding factor 2, Telomeric repeat binding protein 2, telomeric repeat-binding factor 2, TERF 2, TERF2, TRBF 2, TRBF2, TRF 2, TRF2, TTAGGG repeat binding factor 2, TTAGGG repeat-binding factor 2
Símbolos de los genes TERF2
Especie del huésped Mouse
Inmunógeno This TRF-2 Antibody (4A794.15) was developed against Baculovirus expressed whole length TRF2 protein, used for immunizing mice and splenocytes used to generate the hybridoma clone (NP_005643).
Peso molecular del antígeno 59.6 kDa
Método de purificación Protein G purified
Cantidad 0.025 mg
Estado normativo RUO
Disciplina de investigación Cancer, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Transcription Factors and Regulators
Primario o secundario Primary
ID de gen (Entrez) 7014
Especificidad de la prueba The TRF2 antibody recognizes full-length TRF2 as well as TRF2 forms lacking both the N-terminal basic domain (B) and the telobox Myb-like C-terminal DNA-binding domain (M). TRF2 forms missing the B and M domains are often referred to as mutant TRF2. Although the exact epitope recognized by the TRF2 antibody has not been mapped, the scientific literature indicates it is in the D or L domain, but not in the B or M domain.
Especies diana Human, Mouse, Rat, Deer, Marsupial
Contenido y almacenamiento Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG1 κ
Mostrar más Mostrar menos

For Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.