missing translation for 'onlineSavingsMsg'
Learn More

Transferrin Antibody (8B9), DyLight 405, Novus Biologicals™

Código de producto. 18758360 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.2 mL
Tamaño de la unidad:
0.20 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18758360 0.2 mL 0.20 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18758360 Proveedor Novus Biologicals N.º de proveedor NB1107904V

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Transferrin Monoclonal antibody specifically detects Transferrin in Human samples. It is validated for ELISA, Immunoassay, Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Transferrin
Aplicaciones ELISA, Immunoassay, Sandwich ELISA
Clasificación Monoclonal
Clon 8B9
Conjugado DyLight 405
Formulación 50mM Sodium Borate
Alias de gen Beta-1 metal-binding globulin, DKFZp781D0156, EC 3.4.21, PRO1557, PRO2086, serotransferrin, Siderophilin, transferrin
Especie del huésped Mouse
Inmunógeno Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Transferrin purified from pooled human plasma.
Método de purificación Protein A purified
Cantidad 0.2 mL
Estado normativo RUO
Disciplina de investigación Apoptosis, Cancer, Cellular Markers, Golgi Apparatus Markers, Loading Controls, Stem Cell Markers
Primario o secundario Primary
ID de gen (Entrez) 7018
Especies diana Human
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.