missing translation for 'onlineSavingsMsg'
Learn More

TRAIP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

280.00€ - 540.75€

Especificaciones

Antígeno TRAIP
Dilución Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Aplicaciones Immunohistochemistry, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Conjugado Unconjugated
Ver más especificaciones

Productos 2
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
18660788
Ver documentos
Novus Biologicals
NBP2-48655-25ul
25 μL
280.00€
25 microlitros
Ver productos alternativos
Ahorre hasta 
»
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
18609947
Ver documentos
Novus Biologicals
NBP2-48655
0.1 mL
ONLINE EXCLUSIVE PRICE
572.00€
540.75€ / 0.10 ml
Ahorro 31.25€
5% Descuento
Ver productos alternativos
Ahorre hasta 
»
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Descripción

Descripción

TRAIP Polyclonal antibody specifically detects TRAIP in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Especificaciones

Especificaciones

TRAIP
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Zinc Finger
PBS (pH 7.2), 40% Glycerol
10293
IgG
Immunogen affinity purified
Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Polyclonal
Purified
RUO
Human
RNF206TRIPRING finger protein 206, TRAF interacting protein, TRAF-interacting protein
This antibody was developed against a recombinant protein corresponding to amino acids: LNLPPVASETVDRLVLESPAPVEVNLKLRRPSFRDDIDLNATFDVDTPPARPSSSQHGYYEKLCLEKSHSPIQDVPKKICKGPRKESQLSL
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.