missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
TRAILR2/TNFRSF10B Monoclonal antibody specifically detects TRAILR2/TNFRSF10B in Human samples. It is validated for Flow Cytometry, CyTOF
Especificaciones
Especificaciones
| Antígeno | TRAILR2/TNFRSF10B |
| Aplicaciones | Flow Cytometry, CyTOF |
| Clasificación | Monoclonal |
| Clon | 71908 |
| Conjugado | HRP |
| Dilución | Flow Cytometry, CyTOF-ready |
| Formulación | PBS |
| Alias de gen | CD262, CD262 antigen, death domain containing receptor for TRAIL/Apo-2L, Death receptor 5, DR5TRICK2B, Fas-like protein, KILLER/DR5, KILLERapoptosis inducing protein TRICK2A/2B, TNF-related apoptosis-inducing ligand receptor 2, TRAIL receptor 2, TRAIL-R2apoptosis inducing receptor TRAIL-R2, TRAILR2cytotoxic TRAIL receptor-2, TRICK2, TRICK2A, TRICKB, tumor necrosis factor receptor superfamily member 10B, tumor necrosis factor receptor superfamily, member 10b, tumor necrosis factor receptor-like protein ZTNFR9, ZTNFR9 |
| Especie del huésped | Mouse |
| Inmunógeno | Mouse myeloma cell line NS0-derived recombinant human TRAIL R2/TNFRSF10B, Ile56-Glu182, Accession # O14763 |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?