missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
TRAF-6 Polyclonal specifically detects TRAF-6 in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Especificaciones
Especificaciones
| Antígeno | TRAF-6 |
| Aplicaciones | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Dilución | Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000 |
| Formulación | Whole antisera with 0.05% Sodium Azide |
| Alias de gen | E3 ubiquitin-protein ligase TRAF6, EC 6.3.2.-, Interleukin-1 signal transducer, RING finger protein 85, RNF85MGC:3310, TNF receptor-associated factor 6 |
| Símbolos de los genes | TRAF6 |
| Especie del huésped | Rabbit |
| Inmunógeno | A synthetic peptide corresponding to amino acids 451-469 (DAKPELLAFQRPTIPRNPK) of human TRAF6 was used as immunogen, GenBank no. NP_665802.1. |
| Vis mere |
For Research Use Only
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?