missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TOMM20 Polyclonal antibody specifically detects TOMM20 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antígeno | TOMM20 |
| Aplicaciones | Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Dilución | Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200, Immunoprecipitation 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Formulación | PBS (pH 7.3), 50% glycerol |
| Alias de gen | KIAA0016Outer mitochondrial membrane receptor Tom20, MAS20, MGC117367, Mitochondrial 20 kDa outer membrane protein, mitochondrial import receptor subunit TOM20 homolog, MOM19, TOM20, translocase of outer mitochondrial membrane 20 homolog (yeast), translocase of outer mitochondrial membrane 20 homolog type II |
| Especie del huésped | Rabbit |
| Inmunógeno | Recombinant fusion protein containing a sequence corresponding to amino acids 1-145 of human TOMM20 (NP_055580.1). MVGRNSAIAAGVCGALFIGYCIYFDRKRRSDPNFKNRLRERRKKQKLAKERAGLSKLPDLKDAEAVQKFFLEEIQLGEELLAQGEYEKGVDHLTNAIAVCGQPQQLLQVLQQTLPPPVFQMLLTKLPTISQRIVSAQSLAEDDVE |
| Método de purificación | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?