missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNF-alpha Antibody (CDP 571 (Humicade)) - Azide and BSA Free, Novus Biologicals™
Human Monoclonal Antibody
Marca: Novus Biologicals NBP2-75908-0.2MG
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
TNF-alpha Monoclonal specifically detects TNF-alpha in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Neutralization.
Especificaciones
| TNF-alpha | |
| Monoclonal | |
| Unconjugated | |
| PBS with 0.02% Proclin 300 | |
| TNF | |
| Human TNF-alpha | |
| Primary | |
| The antibody binds to human TNF-alpha with a Kd of 0.087 nM. | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
| Western Blot, ELISA, Immunohistochemistry, Neutralization | |
| CDP 571 (Humicade) | |
| Western Blot, ELISA, Immunohistochemistry, Neutralization | |
| APC1 protein, Cachectin, DIF, TNF, monocyte-derived, TNF-a, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
| Human | |
| 0.2 mg | |
| 7124 | |
| Human | |
| IgG4 κ |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido