missing translation for 'onlineSavingsMsg'
Learn More

TLR4 Antibody (76B357.1) - Azide Free, Novus Biologicals™

Código de producto. 18719353 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.025 mg
0.1 mg
Tamaño de la unidad:
0.10 miligramo
25 microgramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18719353 0.1 mg 0.10 miligramo
18137063 0.025 mg 25 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18719353 Proveedor Novus Biologicals N.º de proveedor NBP227149

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody has been used in 25 publications

TLR4 Monoclonal specifically detects TLR4 in Human, Mouse, Rat, Porcine, Bovine, Mammal samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Cell Surface), Flow (Intracellular), CyTOF-ready, Knockout Validated, Knockdown Validated, Block/Neutralize.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno TLR4
Aplicaciones Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), KnockDown, Neutralization
Clasificación Monoclonal
Clon 76B357.1
Concentración 1.0 mg/ml
Conjugado Unconjugated
Dilución Western Blot 1-3 ug/ml, Flow Cytometry 1ul/1 million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 5ug/ml, Immunohistochemistry-Frozen 1:500, Flow (Cell Surface) reported in multiple pieces of scientific literature, Flow (Intracellular) reported in scientific literature (PMID 18577706), Chromatin Immunoprecipitation (ChIP) 1:10-1:500, CyTOF-ready, Knockout Validated, KnockDown Validated, Block/Neutralize reported in scientific literature (PMID 26063617)
N.º de referencia del gen O00206
Alias de gen ARMD10, CD284, CD284 antigen, hTollhomolog of Drosophila toll, TOLL, toll-like receptor 4
Símbolos de los genes TLR4
Especie del huésped Mouse
Inmunógeno This antibody was developed against a portion of amino acids 100-200 of human TLR4 (NP_612564).
Método de purificación Protein G purified
Cantidad 0.1 mg
Estado normativo RUO
Disciplina de investigación Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Toll Like Receptors
Primario o secundario Primary
ID de gen (Entrez) 7099
Especificidad de la prueba The amino acid sequence used has 100% identity in human, chimp, baboon, Chinese hamster, and rat; 93% identity in bovine and pig; 85% identity in sheep and 78% identity in cat, horse and mouse.
Especies diana Human, Mouse, Rat, Pig, Bovine, Mammalia
Contenido y almacenamiento Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG2b κ
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.