Learn More
Novus Biologicals™ TLR3 Antibody (CNTO5429) - Humanized, IgG2SA, Novus Biologicals™
Código de producto. 30045998 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
1 mg
100 μg
50 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
50 microgramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
30045998 50 μg 50 microgramos
30047287 100 μg 100 microgramos
30048547 1 mg 1 miligramo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 30045998 Proveedor Novus Biologicals™ N.º de proveedor NBP32819450ug
Human Monoclonal Antibody
TLR3 Monoclonal antibody specifically detects TLR3 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno TLR3
Aplicaciones Flow Cytometry, ELISA, Functional Assay
Clasificación Monoclonal
Clon CNTO5429
Conjugado Unconjugated
Formulación Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
N.º de referencia del gen O15455
Alias de gen CD283, CD283 antigen, toll-like receptor 3
Especie del huésped Human
Inmunógeno TLR3/ CD283
Método de purificación Protein A purified
Cantidad 50 μg
Estado normativo RUO
Disciplina de investigación Cytokine Research, Immune System Diseases, Immunology, Infections (Virus Bacteria and Parasites), Innate Immunity, Necroptosis, Signal Transduction, Toll Like Receptors, Virology Bacteria and Parasites
Primario o secundario Primary
ID de gen (Entrez) 7098
Reconstitución Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Especies diana Human
Contenido y almacenamiento Store at -20°C in powder form. Store at -80°C once reconstituted.
Formulario Purified
Isotype IgG2