missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 16 publications
223.65€ - 537.60€
Especificaciones
| Antígeno | TLR3 |
|---|---|
| Concentración | 1.0 mg/mL |
| Dilución | Western Blot 1-3 ug/ml, Flow Cytometry 1 ug/10^6 cells, Immunohistochemistry 1:20-1:1000, Immunohistochemistry-Paraffin 1:100. Use reported in scientific literature (Patole et al (2005)), Immunohistochemistry-Frozen 1:20-1:1000. Use reported in scientific literature (Pawar et al (2005)), Dot Blot reported in scientific literature (PMID 27248820), Flow (Cell Surface) reported in scientific literature (Pawar et al (2006)), Flow (Intracellular) 1 ug/1 million cells, KnockDown Validated |
| Aplicaciones | Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Dot Blot, KnockDown |
| Clasificación | Polyclonal |
| Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
|---|---|---|---|---|---|---|---|---|---|
| Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
|
18064005
|
Novus Biologicals
NB100-56571SS |
0.025 mg |
237.00€ 223.65€ / 25 microgramos Ahorro 13.35€ 5% Descuento |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
|
18729313
|
Novus Biologicals
NB100-56571 |
0.1 mg |
569.00€ 537.60€ / 0.10 miligramo Ahorro 31.40€ 5% Descuento |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
Descripción
TLR3 Polyclonal specifically detects TLR3 in Human, Mouse, Porcine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Dot Blot, Flow (Cell Surface), Flow (Intracellular), Knockdown Validated.Especificaciones
| TLR3 | |
| Western Blot 1-3 ug/ml, Flow Cytometry 1 ug/10^6 cells, Immunohistochemistry 1:20-1:1000, Immunohistochemistry-Paraffin 1:100. Use reported in scientific literature (Patole et al (2005)), Immunohistochemistry-Frozen 1:20-1:1000. Use reported in scientific literature (Pawar et al (2005)), Dot Blot reported in scientific literature (PMID 27248820), Flow (Cell Surface) reported in scientific literature (Pawar et al (2006)), Flow (Intracellular) 1 ug/1 million cells, KnockDown Validated | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Pig | |
| O15455 | |
| 7098 | |
| A mix of synthetic peptides corresponding to amino acids 135-150 (SIHKIKSNPFKNQKNL), 828-844 (CRRFKVHHAVQQAIEQN), and 876-891 (CILNWPVQKERINAFH) of mouse TLR3. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 104 kDa |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Dot Blot, KnockDown | |
| Unconjugated | |
| Rabbit | |
| Immune System Diseases, Immunology, Innate Immunity, Signal Transduction, Toll Like Receptors | |
| PBS with 0.05% Sodium Azide | |
| CD283, CD283 antigen, toll-like receptor 3 | |
| TLR3 | |
| IgG | |
| Protein G purified | |
| Mouse and Human. Predicted to cross-react with Rat. Porcine data from customer review. |
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto