Descripción
TLR2 Monoclonal antibody specifically detects TLR2 in Human samples. It is validated for Western Blot
Especificaciones
Especificaciones
| Antígeno | TLR2 |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | CL10552 |
| Conjugado | mFluor Violet 610 SE |
| Formulación | 50mM Sodium Borate |
| Alias de gen | CD282 antigen, TIL4CD282, toll/interleukin 1 receptor-like 4, Toll/interleukin-1 receptor-like protein 4, toll-like receptor 2 |
| Especie del huésped | Mouse |
| Inmunógeno | This antibody was developed using a synthetic peptide derived from O60603, with the exact immunogen sequence remaining proprietary. |
| Método de purificación | Protein A purified |
| Mostrar más |