Learn More
Eurofins DiscoverX TLK2 Protein, Active
Código de producto. 17300644
Encompass_Preferred
Click to view available options
Cantidad:
10 μg
250 μg
Tamaño de la unidad:
10 microgramos
250 microgramos
Código de producto. 17300644
Marca: Eurofins DiscoverX 14739
Active, N-terminal 6His-tagged, recombinant, 388-end, human TLK2, for use in Kinase Assays.
The Tousled-like kinases, first described in Arabidopsis, are nuclear serine/threonine kinases that are potentially involved in the regulation of chromatin assembly. Rapidly and transiently inhibited by phosphorylation following the generation of DNA double-stranded breaks during S- phase. This is cell cycle checkpoint and ATM-pathway dependent and appears to regulate processes involved in chromatin assembly.
GenBank BC044925
Especificaciones
Formulación mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution.
Peso molecular 48.8 kDa
Cantidad 10 μg
Especie Human
Recombinante Recombinant
Etiqueta de proteína N-terminal 6His
Sistema de expresión Recombinant enzyme expressed in Sf21 insect cells
Forma de proteína Truncated
Grado de pureza o calidad ≥60%
Expresión TLK2, active