missing translation for 'onlineSavingsMsg'
Learn More

TLE3 Antibody (CL3573) [PE], Novus Biologicals™

Product Code. 30303856 Shop All Bio Techne Products
Change view
Click to view available options
Cantidad:
0.1 mL
Unit Size:
0.10 ml
Product Code. Cantidad unitSize
30303856 0.1 mL 0.10 ml
1 options
This item is not returnable. View return policy

Product Code. 30303856

Brand: Novus Biologicals NBP261621PE

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TLE3 Monoclonal antibody specifically detects TLE3 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antígeno TLE3
Aplicaciones Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown, Immunocytochemistry/Immunofluorescence
Clasificación Monoclonal
Clon CL3573
Conjugado PE
Formulación PBS
Alias de gen ESG3Enhancer of split groucho-like protein 3, ESGenhancer of split groucho 3, GRG3, HsT18976, KIAA1547FLJ39460, transducin-like enhancer of split 3 (E(sp1) homolog, Drosophila), transducin-like enhancer of split 3, homolog of Drosophila E(sp1), transducin-like enhancer protein 3
Especie del huésped Mouse
Inmunógeno This antibody was developed using a recombinant protein derived from Q04726, with the exact immunogen sequence remaining proprietary.
Método de purificación Protein A purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Signal Transduction
Primario o secundario Primary
ID de gen (Entrez) 7090
Especies diana Human
Contenido y almacenamiento Store at 4⪚C in the dark.
Formulario Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.