Learn More
DiscoverX TIP-39
Código de producto. 17332409
Encompass
Change view
Click to view available options
:
1 mg
Código de producto.
17332409 1 mg
1 options
Código de producto. 17332409
Marca: DiscoverX 921071
TIP-39 is an agonist for PTH2R. Provided as 1mg of dry powder.
Tuberoinfundibular peptide of 39 residues (TIP 39) is a neuropeptide protein related to parathyroid hormone that is a potent and selective agonist of parathyroid hormone 2 receptor (PTH2R), resulting in adenyl cyclase activation and elevation of intracellular calcium levels. Also plays a role in protein kinase C beta activation, beta-arrestin recruitment, PTH2R internalization, cell proliferation inhibition, and possibly spermatogenesis and nociceptive circuit activation. Has little influence on PTH1R. Provided as 1mg of dry powder.
Especificaciones
Cantidad 1 mg
Tipo de producto Control Ligands