missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Tie-2 Antibody Pair [HRP]

Código de producto. 18639540 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
15 placas
5 placas
Tamaño de la unidad:
paquete de 15
paquete de 5
Código de producto. Cantidad unitSize
18639540 5 placas paquete de 5
18244346 15 placas paquete de 15
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Product Code. 18639540

Brand: Novus Biologicals™ NBP2795185Plates

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Polyclonal/Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes an antibody specific for Human Tie-2.
TRUSTED_SUSTAINABILITY

Specifications

Intervalo del ensayo 78.125 pg/mL
Sensibilidad del ensayo 78.125-5000 pg/mL
ensayo Sandwich ELISA
Conjugado HRP
Tipo de producto Antibody Pairs
Especificidad This solid phase sandwich ELISA utilizes an antibody specific for Human Tie-2.
Especies diana Human
Para utilizar con (aplicación) Sandwich ELISA
Alias de gen angiopoietin-1 receptor, CD202B, CD202b antigen, EC 2.7.10, EC 2.7.10.1, hTIE2, p140 TEK, soluble TIE2 variant 1, soluble TIE2 variant 2, TEK tyrosine kinase, endothelial, TIE-2, TIE2CD202b, Tunica interna endothelial cell kinase, Tyrosine-protein kinase receptor TEK, Tyrosine-protein kinase receptor TIE-2, venous malformations, multiple cutaneous and mucosal, VMCM, VMCM1
ID de gen (Entrez) 7010
Símbolo de gen TEK
Especie del huésped Rabbit
Contenido del kit
  • Standard: Each vial contains 245 ng of recombinant Human Tie-2.
  • Capture Antibody: 1.5 mg/mL of rabbit anti-Human Tie-2 monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.2 mg/mL of rabbit anti-Human Tie-2 polyclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4°C).
  • Cantidad 5 placas
    Estado normativo RUO
    Disciplina de investigación Angiogenesis, Cancer, Phospho Specific, Protein Kinase
    Requisitos de almacenamiento Storage is content dependent.
    Objetivo Tie-2
    Técnica 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 ug/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20°C to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 5000 pg/mL is recommended.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.