missing translation for 'onlineSavingsMsg'
Learn More

Thyroglobulin Antibody (TGB/1970R), DyLight 550, Novus Biologicals™

Código de producto. 18577103 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18577103 100 μg 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Product Code. 18577103 Supplier Novus Biologicals Supplier No. NBP308265R

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Thyroglobulin Monoclonal antibody specifically detects Thyroglobulin in Human, Mouse (Negative), Rat (Negative) samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Thyroglobulin
Aplicaciones Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon TGB/1970R
Conjugado DyLight 550
Formulación 50mM Sodium Borate
Alias de gen AITD3TGN, TDH3, Tg, thyroglobulin
Especie del huésped Rabbit
Inmunógeno Recombinant full-length human Thyroglobulin protein (Uniprot: P01266)
Método de purificación Protein A or G purified
Cantidad 100 μg
Estado normativo RUO
Disciplina de investigación Cancer, Cell Biology
Primario o secundario Primary
ID de gen (Entrez) 7038
Especies diana Human, Mouse (Negative), Rat (Negative)
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.