missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Thrombomodulin/BDCA-3 Monoclonal antibody specifically detects Thrombomodulin/BDCA-3 in Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry
Especificaciones
Especificaciones
| Antígeno | Thrombomodulin/BDCA-3 |
| Aplicaciones | Western Blot, Flow Cytometry, Immunohistochemistry |
| Clasificación | Monoclonal |
| Clon | SR1034 |
| Conjugado | Unconjugated |
| Dilución | Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200 |
| Formulación | PBS, pH 7.4, 150mM NaCl, 50% glycerol. |
| Alias de gen | BDCA3, CD141, CD141 antigen, fetomodulin, thrombomodulin, TM |
| Especie del huésped | Rabbit |
| Inmunógeno | A synthesized peptide derived from human Thrombomodulin/BDCA-3 (Uniprot #: P15306) |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?