missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGF-beta 3 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Marca: R&D Systems MAB643-500
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
TGF-beta 3 Monoclonal specifically detects TGF-beta 3 in Multi-Species samples. It is validated for Western Blot, Immunohistochemistry, ELISA Capture (Matched Antibody Pair).
Especificaciones
| TGF-beta 3 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ARVD, ARVD1, FLJ16571, LDS5, RNHF, TGFB3, TGFbeta 3, TGF-beta3, TGF-beta-3, transforming growth factor beta-3, transforming growth factor, beta 3 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7043 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Immunohistochemistry, ELISA | |
| 44922 | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| NP_003230.1 | |
| TGFB3 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human TGF-beta 3 Ala301-Ser412 (Tyr340Phe) Accession # NP_003230.1 | |
| 500 μg | |
| Primary | |
| Detects TGF-beta 3 from multiple species in ELISAs and Western blots. In ELISAs, does not cross-react with recombinant human (rh) TGF-beta 1or rhTGF-beta 2. | |
| Multi-species | |
| Supernatant |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido