missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
TEV Cleavage Site Polyclonal specifically detects TEV Cleavage Site in Non-species specific samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones
Especificaciones
| Antígeno | TEV Cleavage Site |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.5 mg/mL |
| Conjugado | Unconjugated |
| Dilución | Western Blot 0.5 ug/ml |
| Formulación | BBS (pH 8.4) with No Preservative |
| Especie del huésped | Rabbit |
| Inmunógeno | Peptide representing the cleavage site recognized by TEV enzyme |
| Método de purificación | Affinity Purified |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
¿Detecta una oportunidad de mejora?