Descripción
TET2 Monoclonal antibody specifically detects TET2 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Especificaciones
Especificaciones
| Antígeno | TET2 |
| Aplicaciones | Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence |
| Clasificación | Monoclonal |
| Clon | CL6873 |
| Conjugado | PE |
| Formulación | PBS |
| Alias de gen | FLJ20032, MGC125715, tet methylcytosine dioxygenase 2 |
| Especie del huésped | Mouse |
| Inmunógeno | This antibody was developed using a synthetic peptide derived from Q6N021, with the exact immunogen sequence remaining proprietary. |
| Método de purificación | Protein A purified |
| Mostrar más |