missing translation for 'onlineSavingsMsg'
Learn More

TET2 Antibody (CL6873) [Allophycocyanin], Novus Biologicals™

Código de producto. 30305096 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
Código de producto. Cantidad unitSize
30305096 0.1 mL 0.10 ml
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 30305096

Marca: Novus Biologicals NBP276510APC

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TET2 Monoclonal antibody specifically detects TET2 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antígeno TET2
Aplicaciones Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
Clasificación Monoclonal
Clon CL6873
Conjugado APC
Formulación PBS
Alias de gen FLJ20032, MGC125715, tet methylcytosine dioxygenase 2
Especie del huésped Mouse
Inmunógeno This antibody was developed using a synthetic peptide derived from Q6N021, with the exact immunogen sequence remaining proprietary.
Método de purificación Protein A purified
Cantidad 0.1 mL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 54790
Especies diana Human
Contenido y almacenamiento Store at 4⪚C in the dark.
Formulario Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.