missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TESK1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA5121550
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
TESK1 serine/threonine kinase or testis-specific kinase 1 contains an N-terminal protein kinase domain and a C-terminal proline-rich domain which is most closely related to those of the LIM motif-containing protein kinases (LIMKs). TESK1 protein can phosphorylate myelin basic protein and histone in vitro and plays an important role during and after the meiotic phase of spermatogenesis. TESK1 is mainly expressed in testicular germ cells.
Especificaciones
| TESK1 | |
| Polyclonal | |
| Unconjugated | |
| TESK1 | |
| AI326901; dual specificity testis-specific protein kinase 1; Tesk1; Testicular protein kinase 1; testis specific protein kinase 1; testis-specific kinase 1 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 21754, 29460, 7016 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| O70146, Q15569, Q63572 | |
| TESK1 | |
| Phe57-Ile314 (Accession Q15569), with N-terminal His Tag. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido