missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TEM8/ANTXR1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP1-77233-0.025mg
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
TEM8/ANTXR1 Polyclonal specifically detects TEM8/ANTXR1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Especificaciones
| TEM8/ANTXR1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| anthrax toxin receptor 1, ATR2310008J16Rik, FLJ10601, FLJ11298, FLJ21776, TEM82810405N18Rik, Tumor endothelial marker 8 | |
| Rabbit | |
| 45 kDa | |
| 0.025 mg | |
| Primary | |
| Recognizes all three isoforms of ATR. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 0.5-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2 μg/mL, Immunocytochemistry/Immunofluorescence 10 μg/mL, Immunohistochemistry-Paraffin 2 μg/mL | |
| Q9H6X2 | |
| ANTXR1 | |
| Antibody was raised against a peptide corresponding to 13 amino acids near the center of human ATR. The immunogen is located within amino acids 220 - 270 of ATR. Amino Acid Squence: RGNGFRHARNVDR | |
| Ion exchange chromatography | |
| RUO | |
| 84168 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction