Learn More
TEM7/PLXDC1 Antibody (197C193 (IM193)) - BSA Free, Novus Biologicals™
Código de producto. p-4992839 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.025 mg
0.1 mg
Tamaño de la unidad:
0.10 miligramo
25 microgramos
18064084 0.025 mg 25 microgramos
18746193 0.1 mg 0.10 miligramo
2 options
Código de producto. 18064084
Marca: Novus Biologicals NB10056557SS
Mouse Monoclonal Antibody has been used in 8 publications
TEM7/PLXDC1 Monoclonal specifically detects TEM7/PLXDC1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno TEM7/PLXDC1
Aplicaciones Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon 197C193 (IM193)
Concentración 1.0 mg/ml
Conjugado Unconjugated
Dilución Western Blot, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500. Use reported in scientific literature (Meng et al (2007)), Immunoprecipitation 1:10-1:500. Use reported in scientific literature (Nanda et al (2004)), Immunohistochemistry-Paraffin 2-5 ug/ml, Immunohistochemistry-Frozen reported in scientific literature (Lee et al (2006))
N.º de referencia del gen Q8IUK5
Alias de gen FLJ45632, plexin domain containing 1,2410003I07Rik, plexin domain-containing protein 1, TEM3FLJ36270, TEM7DKFZp686F0937, Tumor endothelial marker 3, Tumor endothelial marker 7
Símbolos de los genes PLXDC1
Especie del huésped Mouse
Inmunógeno Amino acids 409-425 (LQNNLSPKTKGTPVHLG) of human TEM7 were used to develop this monoclonal antibody.
Método de purificación Protein G purified
Cantidad 0.025 mg
Estado normativo RUO
Disciplina de investigación Angiogenesis
Primario o secundario Primary
ID de gen (Entrez) 57125
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG1
Mostrar más Mostrar menos
For Research Use Only