Learn More
TEL2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
292.00€ - 556.50€
Especificaciones
Antígeno TEL2
Dilución Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200
Aplicaciones Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Conjugado Unconjugated
Ver más especificaciones
Productos 2
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
18437400
Ver documentos
Novus Biologicals
NBP1-81761-25ul
25 μL
292.00€
25 microlitros
Ver productos alternativos
Ahorre hasta 
»
18229425
Ver documentos
Novus Biologicals
NBP1-81761
0.1 mL
ONLINE EXCLUSIVE PRICE
589.00€
556.50€ / 0.10 ml
Ahorro 32.50€
5% Descuento
Ver productos alternativos
Ahorre hasta 
»
Descripción
Descripción
TEL2 Polyclonal specifically detects TEL2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Especificaciones
Especificaciones
TEL2
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
RUO
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
9894
This antibody was developed against Recombinant Protein corresponding to amino acids:SKAVLICLAQLGEPELRDSRDELLASMMAGVKCRLDSSLPPVRRLGMIVAEVVSARIHPEGPPLKFQYEEDELSLELLALASPQPAGDGASEAGTSLVPATA
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200
Polyclonal
Rabbit
Human
CLK2, DKFZp434A073, EC 1.1.1.27, EC 2.7.11.1, HCLK2, KIAA0683FLJ10924, Protein clk-2 homolog, TEL2, TEL2, telomere maintenance 2, homolog (S. cerevisiae), telomere length regulation protein TEL2 homolog
TELO2
IgG
Affinity Purified
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Documentos
Product Certifications
For Research Use Only