missing translation for 'onlineSavingsMsg'
Learn More

TEAD2 Antibody, Novus Biologicals™

Product Code. p-200073563 Shop All Bio Techne Products
Change view
Click to view available options
Cantidad:
100 μL
25 μL
Unit Size:
100 microlitros
25 microlitros
Product Code. Cantidad unitSize
18282353 100 μL 100 microlitros
18631568 25 μL 25 microlitros
2 options
This item is not returnable. View return policy

Product Code. 18282353

Brand: Novus Biologicals NBP255789

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TEAD2 Polyclonal specifically detects TEAD2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno TEAD2
Aplicaciones Immunocytochemistry, Immunofluorescence
Clasificación Polyclonal
Conjugado Unconjugated
Dilución Immunocytochemistry/Immunofluorescence 1-4 ug/ml
Formulación PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Alias de gen TEA domain family member 2TEF4ETF, TEAD-2, TEF-4, transcriptional enhancer factor TEF-4
Símbolos de los genes TEAD2
Especie del huésped Rabbit
Inmunógeno This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:TARLQLVEFSAFVEPPDAVDSYQRHLFVHISQHCPSPGAPPLESVDV
Método de purificación Affinity Purified
Cantidad 100 μL
Estado normativo RUO
Disciplina de investigación Core ESC Like Genes, Stem Cell Markers
Primario o secundario Primary
ID de gen (Entrez) 8463
Especificidad de la prueba Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Especies diana Human
Contenido y almacenamiento Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.