missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
TDG Monoclonal specifically detects TDG in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Especificaciones
Especificaciones
| Antígeno | TDG |
| Aplicaciones | Western Blot, Immunohistochemistry |
| Clasificación | Monoclonal |
| Clon | OTI3C11 |
| Conjugado | Janelia Fluor 549 |
| Dilución | Western Blot, Immunohistochemistry |
| Formulación | 50mM Sodium Borate with 0.05% Sodium Azide |
| Alias de gen | EC 3.2.2.29, thymine-DNA glycosylaseG/T mismatch-specific thymine DNA glycosylase |
| Símbolos de los genes | TDG |
| Especie del huésped | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?