missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TCR1-N6 Monoclonal specifically detects TCR1-N6 in Goat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
Specifications
| Antígeno | TCR1-N6 |
| Aplicaciones | Flow Cytometry, Immunohistochemistry, Functional Assay |
| Clasificación | Monoclonal |
| Clon | CACTB6A |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Dilución | Flow Cytometry 1:10 - 1:1000 |
| Formulación | Ascites with 0.09% Sodium Azide |
| Especie del huésped | Mouse |
| Inmunógeno | Bovine ConA stimulated mononuclear leukocytes |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?