missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
TCR gamma/delta Monoclonal antibody specifically detects TCR gamma/delta in Human, Primate samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Functional Assay, Flow (Cell Surface)
Especificaciones
Especificaciones
| Antígeno | TCR gamma/delta |
| Aplicaciones | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Functional Assay |
| Clasificación | Monoclonal |
| Clon | B1 |
| Conjugado | PE |
| Formulación | PBS |
| Alias de gen | T cell receptor gamma locus, T-cell antigen receptor, gamma polypeptide, T-cell rearranging gene, gamma, T-cell receptor, gamma cluster, TCRG, TRG |
| Especie del huésped | Mouse |
| Inmunógeno | TCR gamma/delta, the subtype of T cell receptor expressed mainly in epithelial tissues and at the sites of infection. |
| Método de purificación | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?