missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCR beta Antibody (H57-597) - Chimeric - Azide and BSA Free, Novus Biologicals™
Tienda Bio Techne ProductosDescripción
TCR beta Monoclonal antibody specifically detects TCR beta in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Especificaciones
Especificaciones
| Antígeno | TCR beta |
| Aplicaciones | Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation |
| Clasificación | Monoclonal |
| Clon | H57-597 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS |
| Alias de gen | T cell receptor beta chain, TCRB, TCRbeta, Tib |
| Especie del huésped | Rabbit |
| Inmunógeno | Hamsters were immunised with affinity purified DO-11.10 TCR, and produced detectable anti-TCR antibodies which were judged by the ability of the anti-sera to immunoprecipitate TCR from surface labelled DO-11.10 T cell hybridomas. Hamster 57 was selected for hybridisation, and B cell hybridomas were produced using polyethylene glycol fusion of hamster splenic B cells to the mouse myeloma variant. Selected mAb were screened after subsequent culturing for their ability to immunoprecipitate TCR from detergent lysates of surface-radiolabelled murine T cell hybridomas. |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?