missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCF7L1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14519-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This gene encodes a member of the T cell factor/lymphoid enhancer factor family of transcription factors. These transcription factors are activated by beta catenin, mediate the Wnt signaling pathway and are antagonized by the transforming growth factor beta signaling pathway. The encoded protein contains a high mobility group-box DNA binding domain and participates in the regulation of cell cycle genes and cellular senescence.Especificaciones
| TCF7L1 | |
| Polyclonal | |
| Unconjugated | |
| TCF7L1 | |
| HMG box transcription factor 3, TCF 3, TCF3, TCF7L1, Transcription factor 7 like 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 21415, 312451, 83439 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.36 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9HCS4, Q9Z1J1 | |
| Tcf7l1 | |
| TCF7L1 Fusion Protein Ag5983 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido