missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
TCF4 Monoclonal antibody specifically detects TCF4 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin), Protein Array
Especificaciones
Especificaciones
| Antígeno | TCF4 |
| Aplicaciones | Western Blot, ELISA, Immunohistochemistry (Paraffin), Protein Array |
| Clasificación | Monoclonal |
| Clon | TCF4/1705 |
| Conjugado | Alexa Fluor 647 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | BHLHB19, bHLHb19E2-2, Class B basic helix-loop-helix protein 19, Immunoglobulin transcription factor 2, ITF-2, ITF2PTHS, MGC149724, SEF-2, SEF2-1, SEF2-1A, SEF2-1B, SEF2MGC149723, SL3-3 enhancer factor 2, TCF-4, transcription factor 4 |
| Especie del huésped | Mouse |
| Inmunógeno | Recombinant human TCF4 protein fragment (around aa 365-671) (exact sequence is proprietary) (Uniprot: P15884 ) |
| Método de purificación | Protein A or G purified |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?