Learn More
TCF4 Antibody (SR2168), Novus Biologicals™
Código de producto. 18668884 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μg
50 μg
Tamaño de la unidad:
100 microlitros
50 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18668884 100 μg 100 microlitros
18641214 50 μg 50 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18668884 Proveedor Novus Biologicals N.º de proveedor NBP322305100ul
Rabbit Monoclonal Antibody
TCF4 Monoclonal antibody specifically detects TCF4 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, ChIP Assay (ChIP)
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno TCF4
Aplicaciones Western Blot, Flow Cytometry, Immunohistochemistry, ChIP Assay
Clasificación Monoclonal
Clon SR2168
Conjugado Unconjugated
Dilución Western Blot 1:500-1:2000, Flow Cytometry 1:80, Immunohistochemistry 1:50-1:200, Chromatin Immunoprecipitation (ChIP)
Formulación PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Alias de gen BHLHB19, bHLHb19E2-2, Class B basic helix-loop-helix protein 19, Immunoglobulin transcription factor 2, ITF-2, ITF2PTHS, MGC149724, SEF-2, SEF2-1, SEF2-1A, SEF2-1B, SEF2MGC149723, SL3-3 enhancer factor 2, TCF-4, transcription factor 4
Especie del huésped Rabbit
Inmunógeno A synthesized peptide derived from human TCF4 (Uniprot #: P15884)
Método de purificación Affinity purified
Cantidad 100 μg
Estado normativo RUO
Disciplina de investigación Transcription Factors and Regulators
Primario o secundario Primary
ID de gen (Entrez) 6925
Especies diana Human
Contenido y almacenamiento Store at -20° C. Avoid freeze/thaw cycles.
Formulario Purified
Isotype IgG