missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Tbp7 Monoclonal antibody specifically detects Tbp7 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence, Serum
Especificaciones
Especificaciones
| Antígeno | Tbp7 |
| Aplicaciones | Western Blot, Immunofluorescence |
| Clasificación | Monoclonal |
| Clon | SR1808 |
| Conjugado | Unconjugated |
| Dilución | Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence, Serum 1:50-1:200 |
| Formulación | PBS, pH 7.4, 150mM NaCl, 50% glycerol. |
| Alias de gen | MB67 interacting protein, MGC13687,26S protease regulatory subunit 6B, MGC23214, MGC8570, protease 26S subunit 6, proteasome (prosome, macropain) 26S subunit, ATPase, 4,26S proteasome AAA-ATPase subunit RPT3, Proteasome 26S subunit ATPase 4, S6Tat-binding protein 7, TBP-7MB67-interacting protein, TBP7MIP224 |
| Especie del huésped | Rabbit |
| Inmunógeno | A synthesized peptide derived from human Tbp7 (Uniprot #: P43686) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?