missing translation for 'onlineSavingsMsg'
Learn More

TBC1D4 Antibody (OTI5E6), HRP, Novus Biologicals™

Product Code. 18148636 Shop All Bio Techne Products
Change view
Click to view available options
Cantidad:
0.1 mL
Unit Size:
0.10 ml
Product Code. Cantidad unitSize
18148636 0.1 mL 0.10 ml
1 options
This item is not returnable. View return policy

Product Code. 18148636

Brand: Novus Biologicals NBP274462H

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TBC1D4 Monoclonal specifically detects TBC1D4 in Human, Mouse, Rat samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno TBC1D4
Aplicaciones Western Blot
Clasificación Monoclonal
Clon OTI5E6
Conjugado HRP
Dilución Western Blot
Formulación PBS with No Preservative
Alias de gen Akt substrate of 160 kDa, AS160Acrg embryonic lethality minimal region ortholog, DKFZp779C0666, KIAA0603TBC (Tre-2, BUB2, CDC16) domain-containing protein, TBC1 domain family member 4, TBC1 domain family, member 4
Símbolos de los genes TBC1D4
Especie del huésped Mouse
Inmunógeno Full length recombinant protein of human TBC1D4 (NP_0055647) produced in HEK293T cells.
Método de purificación Affinity Purified
Cantidad 0.1 mL
Disciplina de investigación Diabetes Research, Lipid and Metabolism, Phospho Specific
Primario o secundario Primary
ID de gen (Entrez) 9882.0
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.