missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TBC1D10A Polyclonal antibody specifically detects TBC1D10A in Human, Mouse samples. It is validated for Western Blot
Specifications
Specifications
| Antígeno | TBC1D10A |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Biotin |
| Dilución | Western Blot |
| Formulación | PBS |
| Alias de gen | EBP50-PDX interactor of 64 kDa, EPI64, rab27A-GAP-alpha, TBC1 domain family member 10A, TBC1 domain family, member 10A |
| Especie del huésped | Rabbit |
| Inmunógeno | The immunogen recognized by the antibody maps to a region between residue 458 to 508 of human EBP50-PDX Interactor of 64 kDa using the numbering given in entry NP_114143.1. |
| Método de purificación | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?