Learn More
TAZ/WWTR1 Antibody (CL0371) [Janelia Fluor™ 646], Novus Biologicals™
Código de producto. 30292213 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
30292213 0.1 mL 0.10 ml
1 options
Código de producto. 30292213 Proveedor Novus Biologicals N.º de proveedor NBP252926JF646
Mouse Monoclonal Antibody
TAZ/WWTR1 Monoclonal antibody specifically detects TAZ/WWTR1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno TAZ/WWTR1
Aplicaciones Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown, Immunocytochemistry/Immunofluorescence
Clasificación Monoclonal
Clon CL0371
Conjugado Janelia Fluor 646
Formulación 50mM Sodium Borate
Alias de gen FLJ27004, FLJ45718, TAZDKFZp586I1419, transcriptional co-activator with PDZ-binding motif, Transcriptional coactivator with PDZ-binding motif, WW domain containing transcription regulator 1, WW domain-containing transcription regulator protein 1
Especie del huésped Mouse
Inmunógeno This antibody was developed using a recombinant protein derived from Q9GZV5, with the exact immunogen sequence remaining proprietary.
Método de purificación Protein A purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Chromatin Research, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Markers, Transcription Factors and Regulators
Primario o secundario Primary
ID de gen (Entrez) 25937
Especies diana Human
Contenido y almacenamiento Store at 4⪚C in the dark.
Formulario Purified
Isotype IgG1