Learn More
t-Plasminogen Activator/tPA Antibody (32B1), Novus Biologicals™
Código de producto. 30037654 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μL
50 μL
Tamaño de la unidad:
100 microlitros
50 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
30037654 50 μL 50 microlitros
30035926 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 30037654 Proveedor Novus Biologicals N.º de proveedor NBP32637750ul
Rabbit Monoclonal Antibody
t-Plasminogen Activator/tPA Monoclonal antibody specifically detects t-Plasminogen Activator/tPA in Human samples. It is validated for Flow Cytometry, ELISA
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno t-Plasminogen Activator/tPA
Aplicaciones Flow Cytometry, ELISA
Clasificación Monoclonal
Clon 32B1
Conjugado Unconjugated
Dilución Flow Cytometry 1:50-1:200, ELISA
Formulación PBS, pH 7.4, 150mM NaCl and 50% glycerol
Alias de gen Alteplase, DKFZp686I03148, EC 3.4.21, EC 3.4.21.68, plasminogen activator, tissue, plasminogen activator, tissue type, reteplase, tissue plasminogen activator (t-PA), tissue-type plasminogen activator, TPA, T-PA, t-plasminogen activator
Especie del huésped Rabbit
Inmunógeno A synthesized peptide derived from Human t-Plasminogen Activator/tPA [UniProt P00750]
Método de purificación Affinity purified
Cantidad 50 μL
Estado normativo RUO
Disciplina de investigación Cancer, Cardiovascular Biology, Cell Biology, Endocrinology
Primario o secundario Primary
ID de gen (Entrez) 5327
Especies diana Human
Contenido y almacenamiento Store at -20 to -70C. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG