Learn More
T Cell Receptor Gamma/Delta Hamster, anti-Mouse, APC, Clone: GL3, STEMCELL Technologies
Hamster Monoclonal Antibody
Marca:  Stemcell Technologies 60104AZ
Encompass_Preferred
 Ver más versiones de este producto
Código de producto. 17179021
  • 533.25€ / 100 microgramos
Descripción
  • The GL3 antibody reacts with the δ chain of the murine T cell receptor γ/δ (TCRγ/δ or TCR gamma/delta), a subtype of the TCR involved in the recognition of both peptide and lipid antigens
  • TCRγ/δ comprises a heterodimer (∼ 80 kDa in humans) of disulfide-linked γ and δ subunits that associates with CD3 on the cell surface
  • It is a member of the immunoglobulin superfamily
  • TCRγ/δ is expressed on a subpopulation of T cells in the circulation but may be found on up to 50% of the T cells in epithelial cell-rich tissues
  • These cells have roles in oral and tumor-associated tolerance as well as autoimmune disease and have been described as a link between the adaptive and innate immune responses
  • Once activated, they secrete effector cytokines in a subtype- and context-specific manner
  • Most γ/δ T cells are CD4-/CD8-, though some express CD8
  • The GL3 antibody recognizes an epitope in the constant region of the δ chain and can reportedly activate TCRγ/δ+ cells
Especificaciones
Especificaciones
T Cell Receptor Gamma/Delta
Monoclonal
APC
100 μg
Mouse
T Cells, T Cells, Other Subsets
Immunohistochemistry, Flow Cytometry, Immunofluorescence, Functional Assay, Blocking Assay
GL3
Hamster
Primary
IgG2 κ
Certificados
Promociones
Promociones
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.