missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Syntrophin alpha 1 Monoclonal specifically detects Syntrophin alpha 1 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Flow Cytometry.
Specifications
Specifications
| Antígeno | Syntrophin alpha 1 |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | OTI1H10 |
| Conjugado | PerCP |
| Dilución | Flow Cytometry |
| Formulación | PBS with 0.05% Sodium Azide |
| Alias de gen | 59 kDa dystrophin-associated protein A1 acidic component 1, dJ1187J4.5, LQT12, Pro-TGF-alpha cytoplasmic domain-interacting protein 1, SNT1alpha-1-syntrophin, syntrophin, alpha 1 (dystrophin-associated protein A1, 59kD, acidic component), syntrophin, alpha 1 (dystrophin-associated protein A1, 59kDa, acidic component), syntrophin-1, TACIP1acidic alpha 1 syntrophin |
| Símbolos de los genes | SNTA1 |
| Especie del huésped | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?