missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synaptobrevin homolog YKT6 Antibody - Azide and BSA Free, Novus Biologicals™
Tienda Bio Techne ProductosDescripción
Synaptobrevin homolog YKT6 Polyclonal antibody specifically detects Synaptobrevin homolog YKT6 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antígeno | Synaptobrevin homolog YKT6 |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Dilución | Western Blot 1:500-1:2000 |
| Formulación | PBS (pH 7.3), 50% glycerol |
| Alias de gen | EC 2.3.1, EC 2.3.1.-, R-SNARE, S. cerevisiae, homolog of, SNARE protein Ykt6, synaptobrevin homolog YKT6, YKT6 v-SNARE homolog (S. cerevisiae), YKT6 v-SNARE protein |
| Especie del huésped | Rabbit |
| Inmunógeno | A synthetic peptide corresponding to a sequence within amino acids 100-198 of human Synaptobrevin homolog YKT6 (NP_006546.1). EFSKQVDRIDWPVGSPATIHYPALDGHLSRYQNPREADPMTKVQAELDETKIILHNTMESLLERGEKLDDLVSKSEVLGTQSKAFYKTARKQNSCCAIM |
| Método de purificación | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?