missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUMO2/3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10947-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This gene encodes a protein that is a member of the SUMO (small ubiquitin-like modifier) protein family. It functions in a manner similar to ubiquitin in that it is bound to target proteins as part of a post-translational modification system. However, unlike ubiquitin which targets proteins for degradation, this protein is involved in a variety of cellular processes, such as nuclear transport, transcriptional regulation, apoptosis, and protein stability. It is not active until the last two amino acids of the carboxy-terminus have been cleaved off. Numerous pseudogenes have been reported for this gene. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.Especificaciones
SUMO2/3 | |
Polyclonal | |
Unconjugated | |
SUMO2 | |
Sumo2, Sumo3 | |
SUMO2/3 Fusion Protein Ag1388 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P55854, P61956, P61957, P61959, Q5XIF4, Q9Z172 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
170930, 20610, 499417, 6612, 6613, 690244 | |
-20°C | |
Liquid |