missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STMN3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11311-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This gene encodes a protein which is a member of the stathmin protein family. Members of this protein family form a complex with tubulins at a ratio of 2 tubulins for each stathmin protein. Microtubules require the ordered assembly of alpha- and beta-tubulins, and formation of a complex with stathmin disrupts microtubule formation and function. A pseudogene of this gene is located on chromosome 22. Alternative splicing results in multiple transcript variants.Especificaciones
| STMN3 | |
| Polyclonal | |
| Unconjugated | |
| STMN3 | |
| SCG10 like protein, SCLIP, Stathmin 3, stathmin like 3, STMN3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20262, 29246, 50861 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.45 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O70166, Q9JHU6, Q9NZ72 | |
| Stmn3 | |
| STMN3 Fusion Protein Ag1853 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido