missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human STK3 (MST2), GST Tag Recombinant Protein
Click to view available options
Cantidad:
100 μg
1 mg
Tamaño de la unidad:
1 miligramo
100 microgramos
Descripción
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length
STK3 (MST2) is a STE kinase family member that acts upstream of the MAP kinase signaling pathways, and is known to be activated by the proapoptotic stimuli staurosporine and FAS ligand. STK3 is a stress-activated, pro-apoptotic kinase which, following caspase-cleavage, enters the nucleus and induces chromatin condensation followed by internucleosomal DNA fragmentation. It is a key component of the Hippo signaling pathway which plays a pivotal role in organ size control and tumor suppression by restricting proliferation and promoting apoptosis. The core of this pathway is composed of a kinase cascade wherein STK3/MST2 and STK4/MST1, in complex with its regulatory protein SAV1, phosphorylates and activates LATS1/2 in complex with its regulatory protein MOB1, which in turn phosphorylates and inactivates YAP1 oncoprotein and WWTR1/TAZ. Phosphorylation of YAP1 by LATS2 inhibits its translocation into the nucleus to regulate cellular genes important for cell proliferation, cell death, and cell migration. STK3/MST2 and STK4/MST1 are required to repress proliferation of mature hepatocytes, to prevent activation of facultative adult liver stem cells (oval cells), and to inhibit tumor formation.
Especificaciones
Especificaciones
| Número de acceso | Q13188 |
| Concentración | See Label |
| Para utilizar con (aplicación) | Kinase Assay |
| Formulación | 50 mM tris with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5 |
| ID de gen (Entrez) | 6788 |
| Peso molecular | 85.5 kDa |
| Nombre | Human STK3 (MST2), GST Tag |
| Cantidad | 1 mg |
| Estado normativo | RUO - research use only |
| Alias de gen | 0610042I06Rik; KB-1458E12.1; KRS1; Mammalian STE20-like protein kinase 2; mess1; MST; Mst2; MST-2; MST2/C; MST2/N; Mst3; protein kinase homolog; serine/threonine kinase 3; serine/threonine kinase 3 (STE20 homolog, yeast); serine/threonine kinase 3 (Ste20, yeast homolog); serine/threonine-protein kinase 3; Serine/threonine-protein kinase 3 20 kDa subunit; Serine/threonine-protein kinase 3 36 kDa subunit; Serine/threonine-protein kinase Krs-1; Ste20; STE20-like kinase MST2; STK3; wu:fc19e11; zgc:55383 |
| Mostrar más |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido