missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human STK25 (YSK1), GST Tag Recombinant Protein

Código de producto. p-7121915
Change view
Click to view available options
Cantidad:
100 μg
1 mg
Tamaño de la unidad:
1 miligramo
100 microgramos
Código de producto. Cantidad unitSize
15478483 100 μg 100 microgramos
10402313 1 mg 1 miligramo
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15478483

Marca: Invitrogen™ PR6722A

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length

STK25 (YSK1) is a serine/threonine kinase that belongs to the GCK family of kinases. It has been implicated as an upstream regulator of MAP kinase signaling pathways. Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains. The STE group (homologs of yeast Sterile 7, 11, 20 kinases) consists of 50 kinases related to the mitogen-activated protein kinase (MAPK) cascade families (Ste7/MAP2K, Ste11/MAP3K, and Ste20/MAP4K). MAP kinase cascades, consisting of a MAPK and one or more upstream regulatory kinases (MAPKKs) have been best characterized in the yeast pheromone response pathway. Pheromones bind to Ste cell surface receptors and activate yeast MAPK pathway.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso O00506
Concentración See Label
Para utilizar con (aplicación) Kinase Assay
Formulación 50 mM tris with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5
ID de gen (Entrez) 10494
Peso molecular 75.6 kDa
Nombre Human STK25 (YSK1), GST Tag
Cantidad 100 μg
Estado normativo RUO - research use only
Alias de gen 1500019J11Rik; AU018434; C86992; hypothetical protein; serine/threonine kinase 25; serine/threonine kinase 25 (STE20 homolog, yeast); serine/threonine kinase 25 (Ste20, yeast homolog); serine/threonine kinase 25 (yeast); serine/threonine-protein kinase 25; SOK1; SOK-1; Ste20, yeast homolog; ste20/oxidant stress response kinase 1; Ste20-like; ste20-like kinase; sterile 20/oxidant stress-response kinase 1; STK25; STK25 protein; Yeast Sps1/Ste20-related kinase 1; Ysk1
Nombre común STK25
Familia Quinasa STE20 Kinase Family
Conjugado Unconjugated
Especie Human
Recombinante Recombinant
Etiqueta de proteína GST-tag
Secuencia Full length
Contenido y almacenamiento -80°C, Avoid Freeze/Thaw Cycles
Sistema de expresión Insect cells
Formulario Liquid
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.