missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STK11IP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP1-82885-25ul
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
STK11IP Polyclonal specifically detects STK11IP in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifica
| STK11IP | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| KIAA1898LKB1 interacting protein, LIP1serine/threonine kinase 11-interacting protein, LKB1-interacting protein 1, LKB1IPserine/threonine-protein kinase 11-interacting protein, serine/threonine kinase 11 interacting protein, STK11IP1STK11 interacting protein | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 114790 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| STK11IP | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:MCLVVVSDRRLYLLKVTGEMREPPASWLQLTLAVPLQDLSGIELGLAGQSLRLEWAAGAGRCVLLPRDARHCRAFLEELLDVLQSL | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto